Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human alpha-Smooth Muscle Actin His Protein
SDP

Catalog No. NBP222670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222670 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222670 Supplier Novus Biologicals™ Supplier No. NBP222670
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 3-377 of Human alpha-Smooth Muscle Actin The Recombinant Human alpha-Smooth Muscle Actin Protein is derived from E. coli. The Recombinant Human alpha-Smooth Muscle Actin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001604
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol
Gene ID (Entrez) 59
Molecular Weight (g/mol) 44.4kDa
Name Actin Alpha 2 Smooth Muscle Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen ACTA2, 3-377aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMEEEDS TALVCDNGSG LCKAGFAGDD APRAVFPSIV GRPRHQGVMV GMGQKDSYVG DEAQSKRGIL TLKYPIEHGI ITNWDDMEKI WHHSFYNELR VAPEEHPTLL TEAPLNPKAN REKMTQIMFE TFNVPAMYVA IQAVLSLYAS GRTTGIVLDS GDGVTHNVPI YEGYALPHAI MRLDLAGRDL TDYLMKILTE RGYSFVTTAE REIVRDIKEK LCYVALDFEN EMATAASSSS LEKSYELPDG QVITIGNERF RCPETLFQPS FIGMESAGIH ETTYNSIMKC DIDIRKDLYA NNVLSGGTTM YPGIADRMQK EITALAPSTM KIKIIAPPER KYSVWIGGSI LASLSTFQQM WISKQEYDEA GPSIVHRKCF
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.