Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human alpha Endosulfine His Protein
SDP

Catalog No. NBP137076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137076 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137076 Supplier Novus Biologicals™ Supplier No. NBP137076
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-121 of Human alpha Endosulfine The Recombinant Human alpha Endosulfine Protein is derived from E. coli. The Recombinant Human alpha Endosulfine Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number O43768
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris buffer (pH 8.0), 10% glycerol, 0.2 mM PMSF
Gene ID (Entrez) 2029
Molecular Weight (g/mol) M.W. (theoretical): 15.5 kDa
Name alpha Endosulfine Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MSQKQEEENP AEETGEEKQD TQEKEGILPE RAEEAKLKAK YPSLGQKPGG SDFLMKRLQK GQKYFDSGDY NMAKAKMKNK QLPSAGPDKN LVTGDHIPTP QDLPQRKSSL VTSKLAGGQV E
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol ENSA
Cross Reactivity Human
Purity or Quality Grade >90%, by SDS-PAGE
Protein alpha Endosulfine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.