Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human AMSH/STAMBP His Protein
SDP

Catalog No. NBP222825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222825 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222825 Supplier Novus Biologicals™ Supplier No. NBP222825
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-424 of Human AMSH/STAMBP The Recombinant Human AMSH/STAMBP Protein is derived from E. coli. The Recombinant Human AMSH/STAMBP Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_998787
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 10617
Molecular Weight (g/mol) theoretical_molecular_weight: L606
Name AMSH/STAMBP Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen STAMBP, 1-424aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSDHGDV SLPPEDRVRA LSQLGSAVEV NEDIPPRRYF RSGVEIIRMA SIYSEEGNIE HAFILYNKYI TLFIEKLPKH RDYKSAVIPE KKDTVKKLKE IAFPKAEELK AELLKRYTKE YTEYNEEKKK EAEELARNMA IQQELEKEKQ RVAQQKQQQL EQEQFHAFEE MIRNQELEKE RLKIVQEFGK VDPGLGGPLV PDLEKPSLDV FPTLTVSSIQ PSDCHTTVRP AKPPVVDRSL KPGALSNSES IPTIDGLRHV VVPGRLCPQF LQLASANTAR GVETCGILCG KLMRNEFTIT HVLIPKQSAG SDYCNTENEE ELFLIQDQQG LITLGWIHTH PTQTAFLSSV DLHTHCSYQM MLPESVAIVC SPKFQETGFF KLTDHGLEEI SSCRQKGFHP HSKDPPLFCS CSHVTVVDRA VTITDLR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.