Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human AP1AR His Protein
SDP

Catalog No. NBP222677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222677 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222677 Supplier Novus Biologicals™ Supplier No. NBP222677
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-302 of Human AP1AR The Recombinant Human AP1AR Protein is derived from E. coli. The Recombinant Human AP1AR Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_061039
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 55435
Molecular Weight (g/mol) 36.4kDa
Name AP1AR Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen AP1AR, 1-302aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGNCCWTQCF GLLRKEAGRL QRVGGGGGSK YFRTCSRGEH LTIEFENLVE SDEGESPGSS HRPLTEEEIV DLRERHYDSI AEKQKDLDKK IQKELALQEE KLRLEEEALY AAQREAARAA KQRKLLEQER QRIVQQYHPS NNGEYQSSGP EDDFESCLRN MKSQYEVFRS SRLSSDATVL TPNTESSCDL MTKTKSTSGN DDSTSLDLEW EDEEGMNRML PMRERSKTEE DILRAALKYS NKKTGSNPTS ASDDSNGLEW ENDFVSAEMD DNGNSEYSGF VNPVLELSDS GIRHSDTDQQ TR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.