Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Aprataxin His Protein
SDP

Catalog No. NBP140397 Shop All R&D Systems Products
Change view
Click to view available options
0.05mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP140397 0.05mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140397 Supplier Novus Biologicals™ Supplier No. NBP140397
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-168 of Human Aprataxin The Recombinant Human Aprataxin Protein is derived from E. coli. The Recombinant Human Aprataxin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q7Z2E3
For Use With (Application) ELISA, Western Blot
Formulation 20mM Tris-HCl buffer (pH7.5), 20% glycerol, 100mM NaCl, 0.1mM PMSF
Gene ID (Entrez) 54840
Molecular Weight (g/mol) 23.9kDa
Name Aprataxin Protein
Purification Method Protein
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWAGSMQD PKMQVYKDEQ VVVIKDKYPK ARYHWLVLPW TSISSLKAVA REHLELLKHM HTVGEKVIVD FAGSSKLRFR LGYHAIPSMS HVHLHVISQD FDSPCLKNKK HWNSFNTEYF LESQAVIEMV QEAGRVTVRD GMPELLKLPL RCHECQQLLP SIPQLKEHLR KHWTQ
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.