Learn More
Description
Specifications
Specifications
| Accession Number | NP_079088 |
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 20% glycerol, 2mM EDTA |
| Gene ID (Entrez) | 79870 |
| Molecular Weight (g/mol) | 17.7kDa |
| Name | BAALC Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MGCGGSRADA IEPRYYESWT RETESTWLTY TDSDAPPSAA APDSGPEAGG LHSGMLEDGL PSNGVPRSTA PGGIPNPEKK TNCETQCPNP QSLSSGPLTQ KQNGLQTTEA KRDAKRMPAK EVTINVTDSI QQMDRSRRIT KNCVN |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
