Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Bub3 His Protein
SDP

Catalog No. NBP222719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222719 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222719 Supplier Novus Biologicals™ Supplier No. NBP222719
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-328 of Human Bub3 The Recombinant Human Bub3 Protein is derived from E. coli. The Recombinant Human Bub3 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004716
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 9184
Molecular Weight (g/mol) 39.5kDa
Name Bub3 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen BUB3, 1-328aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTGSNEF KLNQPPEDGI SSVKFSPNTS QFLLVSSWDT SVRLYDVPAN SMRLKYQHTG AVLDCAFYDP THAWSGGLDH QLKMHDLNTD QENLVGTHDA PIRCVEYCPE VNVMVTGSWD QTVKLWDPRT PCNAGTFSQP EKVYTLSVSG DRLIVGTAGR RVLVWDLRNM GYVQQRRESS LKYQTRCIRA FPNKQGYVLS SIEGRVAVEY LDPSPEVQKK KYAFKCHRLK ENNIEQIYPV NAISFHNIHN TFATGGSDGF VNIWDPFNKK RLCQFHRYPT SIASLAFSND GTTLAIASSY MYEMDDTEHP EDGIFIRQVT DAETKPKSPC T
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.