Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human BUD31 His Protein
SDP

Catalog No. NBP222720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222720 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222720 Supplier Novus Biologicals™ Supplier No. NBP222720
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-144 of Human BUD31 The Recombinant Human BUD31 Protein is derived from E. coli. The Recombinant Human BUD31 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003901
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 40% glycerol, 2mM DTT
Gene ID (Entrez) 8896
Molecular Weight (g/mol) 19.4kDa
Name BUD31 Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen BUD31, 1-144 aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMPKVKRS RKAPPDGWEL IEPTLDELDQ KMREAETEPH EGKRKVESLW PIFRIHHQKT RYIFDLFYKR KAISRELYEY CIKEGYADKN LIAKWKKQGY ENLCCLRCIQ TRDTNFGTNC ICRVPKSKLE VGRIIECTHC GCRGCSG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.