Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD74 His Protein
SDP

Catalog No. NBP222758 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222758 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222758 Supplier Novus Biologicals™ Supplier No. NBP222758
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 73-232 of Human CD74 The Recombinant Human CD74 Protein is derived from E. coli. The Recombinant Human CD74 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004346
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 972
Molecular Weight (g/mol) 20.6kDa
Name CD74 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CD74, 73-232aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSQQQGRLD KLTVTSQNLQ LENLRMKLPK PPKPVSKMRM ATPLLMQALP MGALPQGPMQ NATKYGNMTE DHVMHLLQNA DPLKVYPPLK GSFPENLRHL KNTMETIDWK VFESWMHHWL LFEMSRHSLE QKPTDAPPKE SLELEDPSSG LGVTKQDLGP VPM
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.