Learn More
Description
Specifications
Specifications
| Accession Number | NP_004633 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 50% glycerol, 2mM DTT |
| Gene ID (Entrez) | 8099 |
| Molecular Weight (g/mol) | 16.6kDa |
| Name | CDK2AP1 Protein |
| Purification Method | Protein |
| Immunogen | MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMSY KPNLAAHMPA AALNAAGSVH SPSTSMATSS QYRQLLSDYG PPSLGYTQGT GNSQVPQSKY AELLAIIEEL GKEIRPTYAG SKSAMERLKR GIIHARGLVR ECLAETERNA RS |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
