Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CEBP alpha His Protein
SDP

Catalog No. NBC118419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118419 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118419 Supplier Novus Biologicals™ Supplier No. NBC118419
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 270-358 of Human CEBP alpha The Recombinant Human CEBP alpha Protein is derived from E. coli. The Recombinant Human CEBP alpha Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P49715
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5) 0.1 M NaCl, 5mM beta-Mercaptoethanol.
Gene ID (Entrez) 1050
Molecular Weight (g/mol) 14.5kDa
Name CEBP alpha Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen CEBP-alpha, (bZIPregion, residues 270-358, His-tag, Human), E.coli (NP_004355).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.