Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Coagulation Factor III/Tissue Factor His Protein
SDP

Catalog No. NBP222785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222785 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222785 Supplier Novus Biologicals™ Supplier No. NBP222785
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 33-251 of Human Coagulation Factor III/Tissue Factor The Recombinant Human Coagulation Factor III/Tissue Factor Protein is derived from E. coli. The Recombinant Human Coagulation Factor III/Tissue Factor Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001984
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 2152
Molecular Weight (g/mol) 25.9kDa
Name Coagulation Factor III/Tissue Factor Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen F3, 33-251aa, Human, His tag, E.coli MSGTTNTVAA YNLTWKSTNF KTILEWEPKP VNQVYTVQIS TKSGDWKSKC FYTTDTECDL TDEIVKDVKQ TYLARVFSYP AGNVESTGSA GEPLYENSPE FTPYLETNLG QPTIQSFEQV GTKVNVTVED ERTLVRRNNT FLSLRDVFGK DLIYTLYYWK SSSSGKKTAK TNTNEFLIDV DKGENYCFSV QAVIPSRTVN RKSTDSPVEC MGQEKGEFRE LEHHHHHH
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.