Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CPSF4 His Protein
SDP

Catalog No. NBP222783 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222783 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222783 Supplier Novus Biologicals™ Supplier No. NBP222783
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-244 of Human CPSF4 The Recombinant Human CPSF4 Protein is derived from E. coli. The Recombinant Human CPSF4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001075028
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 10898
Molecular Weight (g/mol) 29.9kDa
Name CPSF4 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CPSF4, 1-244aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMQEIIAS VDHIKFDLEI AVEQQLGAQP LPFPGMDKSG AAVCEFFLKA ACGKGGMCPF RHISGEKTVV CKHWLRGLCK KGDQCEFLHE YDMTKMPECY FYSKFGECSN KECPFLHIDP ESKIKDCPWY DRGFCKHGPL CRHRHTRRVI CVNYLVGFCP EGPSCKFMHP RFELPMGTTE QPPLPQQTQP PAKQRTPQVI GVMQSQNSSA GNRGPRPLEQ VTCYKCGEKG HYANRCTKGH LAFLSGQ
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.