Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cyclin I His Protein
SDP

Catalog No. NBP222970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222970 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222970 Supplier Novus Biologicals™ Supplier No. NBP222970
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-377 of Human Cyclin I The Recombinant Human Cyclin I Protein is derived from E. coli. The Recombinant Human Cyclin I Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_006826
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 10983
Molecular Weight (g/mol) 44.9kDa
Name Cyclin Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CCN1, 1-377aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMKFPGPL ENQRLSFLLE KAITREAQMW KVNVRKMPSN QNVSPSQRDE VIQWLAKLKY QFNLYPETFA LASSLLDRFL ATVKAHPKYL SCIAISCFFL AAKTVEEDER IPVLKVLARD SFCGCSSSEI LRMERIILDK LNWDLHTATP LDFLHIFHAI AVSTRPQLLF SLPKLSPSQH LAVLTKQLLH CMACNQLLQF RGSMLALAMV SLEMEKLIPD WLSLTIELLQ KAQMDSSQLI HCRELVAHHL STLQSSLPLN SVYVYRPLKH TLVTCDKGVF RLHPSSVPGP DFSKDNSKPE VPVRGTAAFY HHLPAASGCK QTSTKRKVEE MEVDDFYDGI KRLYNEDNVS ENVGSVCGTD LSRQEGHASP CPPLQPVSVM
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.