Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cyclophilin-E His Protein
SDP

Catalog No. NBC118531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118531 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118531 Supplier Novus Biologicals™ Supplier No. NBC118531
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Specific activity is > 210 nmol/min/ug, and is defined as the amount of enzyme that cleaves 1umole of suc-AAPF-pNA per minute at 1C in Tris-Hcl pH8.0 using chymotrypsin.

Specifications

Accession Number NP_006103
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris buffer (pH 8.0)
Gene ID (Entrez) 10450
Molecular Weight (g/mol) 37.5kDa
Name Cyclophilin-E Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1-301aa, Human,His-Tagged, Recombinant, E.coli
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.