Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human DHPS His Protein
SDP

Catalog No. NBP141227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141227 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141227 Supplier Novus Biologicals™ Supplier No. NBP141227
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-369 of Human DHPS The Recombinant Human DHPS Protein is derived from E. coli. The Recombinant Human DHPS Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001921
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl
Gene ID (Entrez) 1725
Molecular Weight (g/mol) M.W. (theoretical): 43.1 kDa
Name DHPS Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MEGSLEREAP AGALAAVLKH SSTLPPESTQ VRGYDFNRGV NYRALLEAFG TTGFQATNFG RAVQQVNAMI EKKLEPLSQD EDQHADLTQS RRPLTSCTIF LGYTSNLISS GIRETIRYLV QHNMVDVLVT TAGGVEEDLI KCLAPTYLGE FSLRGKELRE NGINRIGNLL VPNENYCKFE DWLMPILDQM VMEQNTEGVK WTPSKMIARL GKEINNPESV YYWAQKNHIP VFSPALTDGS LGDMIFFHSY KNPGLVLDIV EDLRLINTQA IFAKCTGMII LGGGVVKHHI ANANLMRNGA DYAVYINTAQ EFDGSDSGAR PDEAVSWGKI RVDAQPVKVY ADASLVFPLL VAETFAQKMD AFMHEKNED
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol DHPS
Cross Reactivity Human
Research Category Cell Cycle and Replication
Purity or Quality Grade >95%, by SDS-PAGE
Protein DHPS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.