Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human DUSP26 His Protein
SDP

Catalog No. NBP223009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223009 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223009 Supplier Novus Biologicals™ Supplier No. NBP223009
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-211 of Human DUSP26 The Recombinant Human DUSP26 Protein is derived from E. coli. The Recombinant Human DUSP26 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_076930
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 78986
Molecular Weight (g/mol) 26.3kDa
Name DUSP26 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen DUSP26, 1-211aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMCPGNWL WASMTFMARF SRSSSRSPVR TRGTLEEMPT VQHPFLNVFE LERLLYTGKT ACNHADEVWP GLYLGDQDMA NNRRELRRLG ITHVLNASHS RWRGTPEAYE GLGIRYLGVE AHDSPAFDMS IHFQTAADFI HRALSQPGGK ILVHCAVGVS RSATLVLAYL MLYHHLTLVE AIKKVKDHRG IIPNRGFLRQ LLALDRRLRQ GLEA
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Neuronal Cell Markers, Neurotransmission, Protein Phosphatase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.