Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EED His Protein
SDP

Catalog No. NBP223020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223020 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223020 Supplier Novus Biologicals™ Supplier No. NBP223020
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-441 of Human EED The Recombinant Human EED Protein is derived from E. coli. The Recombinant Human EED Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003788
For Use With (Application) In vitro Assay, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 8726
Molecular Weight (g/mol) 52.6kDa
Name EED Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen EED, 1-441aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSEREVS TAPAGTDMPA AKKQKLSSDE NSNPDLSGDE NDDAVSIESG TNTERPDTPT NTPNAPGRKS WGKGKWKSKK CKYSFKCVNS LKEDHNQPLF GVQFNWHSKE GDPLVFATVG SNRVTLYECH SQGEIRLLQS YVDADADENF YTCAWTYDSN TSHPLLAVAG SRGIIRIINP ITMQCIKHYV GHGNAINELK FHPRDPNLLL SVSKDHALRL WNIQTDTLVA IFGGVEGHRD EVLSADYDLL GEKIMSCGMD HSLKLWRINS KRMMNAIKES YDYNPNKTNR PFISQKIHFP DFSTRDIHRN YVDCVRWLGD LILSKSCENA IVCWKPGKME DDIDKIKPSE SNVTILGRFD YSQCDIWYMR FSMDFWQKML ALGNQVGKLY VWDLEVEDPH KAKCTTLTHH KCGAAIRQTS FSRDSSILIA VCDDASIWRW DRLR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Purity or Quality Grade >90% SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.