Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human eIF2 alpha/EIF2S1 His Protein
SDP

Catalog No. NBP144467 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144467 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144467 Supplier Novus Biologicals™ Supplier No. NBP144467
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-315 of Human eIF2 alpha/EIF2S1 The Recombinant Human eIF2 alpha/EIF2S1 Protein is derived from E. coli. The Recombinant Human eIF2 alpha/EIF2S1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004085
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 1965
Molecular Weight (g/mol) 38.2kDa
Name eIF2 alpha/EIF2S1 Protein
Purification Method Protein
Quantity 50 μg
Immunogen MGSSHHHHHH SSGLVPRGSH MPGLSCRFYQ HKFPEVEDVV MVNVRSIAEM GAYVSLLEYN NIEGMILLSE LSRRRIRSIN KLIRIGRNEC VVVIRVDKEK GYIDLSKRRV SPEEAIKCED KFTKSKTVYS ILRHVAEVLE YTKDEQLESL FQRTAWVFDD KYKRPGYGAY DAFKHAVSDP SILDSLDLNE DEREVLINNI NRRLTPQAVK IRADIEVACY GYEGIDAVKE ALRAGLNCST ENMPIKINLI APPRYVMTTT TLERTEGLSV LSQAMAVIKE KIEEKRGVFN VQMEPKVVTD TDETELARQM ERLERENAEV DGDDDAEEME AKAED
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.