Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FKBP13/FKBP2 Protein
SDP

Catalog No. NBP141233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141233 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141233 Supplier Novus Biologicals™ Supplier No. NBP141233
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

Specific activity is > 700 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmol of suc-AAPF-pNA per minute at 37C in Tris-HCl pH 8.0 using chymotrypsin.

Specifications

Accession Number NP_001128680
Concentration 1 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1 mM DTT
Gene ID (Entrez) 2286
Molecular Weight (g/mol) M.W. (theoretical): 13.4 kDa
Name FKBP13/FKBP2 Protein
Quantity 0.05 mg
Source E. Coli
Immunogen 22-142aa, MATGAEGKRK LQIGVKKRVD HCPIKSRKGD VLHMHYTGKL EDGTEFDSSL PQNQPFVFSL GTGQVIKGWD QGLLGMCEGE KRKLVIPSEL GYGERGAPPK IPGGATLVFE VELLKIERRT EL
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of protein (determined by LAL method)
Gene Symbol FKBP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE
Protein FKBP13/FKBP2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.