Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FKBP6 His Protein
SDP

Catalog No. NBP144384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144384 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144384 Supplier Novus Biologicals™ Supplier No. NBP144384
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Specific activity is > 190 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAPF-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.

Specifications

Accession Number NP_003593
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol, 1mM EDTA, 50mM NaCl
Gene ID (Entrez) 8468
Molecular Weight (g/mol) 39.3kDa
Name FKBP6 Protein
Purification Method Protein
Quantity 0.05 mg
Source E. Coli
Immunogen 1-327aa, Human, His-tagged, MGSSHHHHHH SSGLVPRGSH MGGSALNQGV LEGDDAPGQS LYERLSQRML DISGDRGVLK DVIREGAGDL VAPDASVLVK YSGYLEHMDR PFDSNYFRKT PRLMKLGEDI TLWGMELGLL SMRRGELARF LFKPNYAYGT LGCPPLIPPN TTVLFEIELL DFLDCAESDK FCALSAEQQD QFPLQKVLKV AATEREFGNY LFRQNRFYDA KVRYKRALLL LRRRSAPPEE QHLVEAAKLP VLLNLSFTYL KLDRPTIALC YGEQALIIDQ KNAKALFRCG QACLLLTEYQ KARDFLVRAQ KEQPFNHDIN NELKKLASCY RDYVDKEKEM WHRMFAPCGD GSTAGES
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.