Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GAMT His Protein
SDP

Catalog No. NBP130287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130287 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130287 Supplier Novus Biologicals™ Supplier No. NBP130287
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-236 of Human GAMT The Recombinant Human GAMT Protein is derived from E. coli. The Recombinant Human GAMT Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000147
Concentration 1mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT,10% glycerol
Gene ID (Entrez) 2593
Molecular Weight (g/mol) 28.4kDa
Name GAMT Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MSAPSATPIF APGENCSPAW GAAPAAYDAA DTHLRILGKP VMERWETPYM HALAAAASSK GGRVLEVGFG MAIAASKVQE APIDEHWIIE CNDGVFQRLR DWAPRQTHKV IPLKGLWEDV APTLPDGHFD GILYDTYPLS EETWHTHQFN FIKNHAFRLL KPGGVLTYCN LTSWGELMKS KYSDITIMFE ETQVPALLEA GFRRENIRTE VMALVPPADC RYYAFPQMIT PLVTKG
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Research Category Proteases & Other Enzymes
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.