Learn More
Description
Specifications
Specifications
| Accession Number | NP_958836 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 2mM DTT, 10% glycerol |
| Gene ID (Entrez) | 8522 |
| Molecular Weight (g/mol) | 11.7kDa |
| Name | GAS7 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MKPGMVPPPP GEESQTVILP PGWQSYLSPQ GRRYYVNTTT NETTWERPSS SPGIPASPGS HRSSLPPTVN GYHASGTPAH PPETAHMSVR KSTGDSQNLG SSSPSKKQSK ENTITINCVT FPHPDTMPEQ QLLKPTEWSY CDYFWADKKD PQGNGTVAGF ELLLQKQLKG KQMQKEMSEF IRERIKIEED YAKNLAKLSQ NSLASQEEGS LGEAWAQVKK SLADEAEVHL KFSAKLHSEV EKPLMNFREN FKKDMKKCDH HIADLRKQLA SRYASVEKAR KALTERQRDL EMKTQQLEIK LSNKTEEDIK KARRKSTQAG DDLMRCVDLY NQAQSKWFEE MVTTTLELER LEVERVEMIR QHLCQYTQLR HETDMFNQST VEPVDQLLRK VDPAKDRELW VREHKTGNIR PVDMEI |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
