Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Geminin His Protein
SDP

Catalog No. NBC118423 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118423 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118423 Supplier Novus Biologicals™ Supplier No. NBC118423
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-209 of Human Geminin The Recombinant Human Geminin Protein is derived from E. coli. The Recombinant Human Geminin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number O75496
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 100 mM NaCl, 10% glycerol
Gene ID (Entrez) 51053
Molecular Weight (g/mol) M.W. (theoretical): 27.6 kDa
Name Geminin Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Geminin, Human, Recombinant, His- tagged, E.coli (NP_056979).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol GMNN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Breast Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers
Purity or Quality Grade >95%, by SDS-PAGE
Protein Geminin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.