Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Glutathione S Transferase kappa 1 Protein
SDP

Catalog No. NBP137083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137083 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137083 Supplier Novus Biologicals™ Supplier No. NBP137083
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-226 of Human Glutathione S Transferase kappa 1 The Recombinant Human Glutathione S Transferase kappa 1 Protein is derived from E. coli. The Recombinant Human Glutathione S Transferase kappa 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_057001
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1 mM DTT
Gene ID (Entrez) 373156
Molecular Weight (g/mol) M.W. (theoretical): 25.5 kDa
Name Glutathione S Transferase kappa 1 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGPLPRTVEL FYDVLSPYSW LGFEILCRYQ NIWNINLQLR PSLITGIMKD SGNKPPGLLP RKGLYMANDL KLLRHHLQIP IHFPKDFLSV MLEKGSLSAM RFLTAVNLEH PEMLEKASRE LWMRVWSRNE DITEPQSILA AAEKAGMSAE QAQGLLEKIA TPKVKNQLKE TTEAACRYGA FGLPITVAHV DGQTHMLFGS DRMELLAHLL GEKWMGPIPP AVNARL
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol GSTK1
Cross Reactivity Human
Purity or Quality Grade >90%, by SDS-PAGE
Protein Glutathione S Transferase kappa 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.