Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GOLM1 His Protein
SDP

Catalog No. NBP223084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223084 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223084 Supplier Novus Biologicals™ Supplier No. NBP223084
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 36-401 of Human GOLM1 The Recombinant Human GOLM1 Protein is derived from E. coli. The Recombinant Human GOLM1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_808800
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 51280
Molecular Weight (g/mol) 44kDa
Name GOLM1 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen GOLM1, 36-401aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSSSRSVDL QTRIMELEGR VRRAAAERGA VELKKNEFQG ELEKQREQLD KIQSSHNFQL ESVNKLYQDE KAVLVNNITT GERLIRVLQD QLKTLQRNYG RLQQDVLQFQ KNQTNLERKF SYDLSQCINQ MKEVKEQCEE RIEEVTKKGN EAVASRDLSE NNDQRQQLQA LSEPQPRLQA AGLPHTEVPQ GKGNVLGNSK SQTPAPSSEV VLDSKRQVEK EETNEIQVVN EEPQRDRLPQ EPGREQVVED RPVGGRGFGG AGELGQTPQV QAALSVSQEN PEMEGPERDQ LVIPDGQEEE QEAAGEGRNQ QKLRGEDDYN MDENEAESET DKQAALAGND RNIDVFNVED QKRDTINLLD QREKRNHTL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.