Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GRP75/HSPA9B/Mortalin His Protein
SDP

Catalog No. NBC118380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118380 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118380 Supplier Novus Biologicals™ Supplier No. NBC118380
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 47-679 of Human GRP75/HSPA9B/Mortalin The Recombinant Human GRP75/HSPA9B/Mortalin Protein is derived from E. coli. The Recombinant Human GRP75/HSPA9B/Mortalin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P38646
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.5mM DTT, 10% glycerol
Gene ID (Entrez) 3313
Molecular Weight (g/mol) 71kDa
Name GRP75/HSPA9B/Mortalin Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen HSPA9, 47-679aa, Human, His-Tagged, Recombinant, E.coli (NP_004125).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.