Learn More
Description
Specifications
Specifications
| Accession Number | NP_005323 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl, 20% glycerol |
| Gene ID (Entrez) | 3050 |
| Molecular Weight (g/mol) | 16.7kDa |
| Name | Hemoglobin zeta Protein |
| Purification Method | Protein |
| Immunogen | MSLTKTERTI IVSMWAKIST QADTIGTETL ERLFLSHPQT KTYFPHFDLH PGSAQLRAHG SKVVAAVGDA VKSIDDIGGA LSKLSELHAY ILRVDPVNFK LLSHCLLVTL AARFPADFTA EAHAAWDKFL SVVSSVLTEK YRLEHHHHHH |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
