Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HIF-1 alpha Protein
SDP

Catalog No. NBC118422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118422 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118422 Supplier Novus Biologicals™ Supplier No. NBC118422
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 530-826 of Human HIF-1 alpha The Recombinant Human HIF-1 alpha Protein is derived from E. coli. The Recombinant Human HIF-1 alpha Protein has been validated for the following applications: SDS-Page, Block/Neutralize.

Specifications

Accession Number Q16665
For Use With (Application) Block/Neutralize, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5) 1mM DTT
Gene ID (Entrez) 3091
Molecular Weight (g/mol) 32.8kDa
Name HIF-1 alpha Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Hif-1a (530-826 aa) Human, Recombinant, E.coli (NP_001521).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.