Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HMGN1/HMG14 His Protein
SDP

Catalog No. NBP141146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141146 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141146 Supplier Novus Biologicals™ Supplier No. NBP141146
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-100 of Human HMGN1/HMG14 The Recombinant Human HMGN1/HMG14 Protein is derived from E. coli. The Recombinant Human HMGN1/HMG14 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004956
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl, 0.1mM PMSF
Gene ID (Entrez) 3150
Molecular Weight (g/mol) 11.7kDa
Name HMGN1/HMG14 Protein
Purification Method Protein
Quantity 0.05 mg
Source E. Coli
Immunogen MGN1, 1-100aa, Human, His tag, E.coli (NP_004956).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.