Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HSF1 His Protein
SDP

Catalog No. NBC118373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118373 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118373 Supplier Novus Biologicals™ Supplier No. NBC118373
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-529 of Human HSF1 The Recombinant Human HSF1 Protein is derived from E. coli. The Recombinant Human HSF1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005517
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 50 mM NaCl, 1 mM DTT
Gene ID (Entrez) 3297
Molecular Weight (g/mol) M.W. (theoretical): 59.4 kDa
Name HSF1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1-529 aa, Human, His-tagged, Recombinant, E.coli
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol HSF1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cancer, DNA Repair, Transcription Factors and Regulators
Purity or Quality Grade >80%, by SDS-PAGE
Protein HSF1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.