Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IGFBP-rp1/IGFBP-7 His Protein
SDP

Catalog No. NBP223142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223142 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223142 Supplier Novus Biologicals™ Supplier No. NBP223142
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 27-282 of Human IGFBP-rp1/IGFBP-7 The Recombinant Human IGFBP-rp1/IGFBP-7 Protein is derived from E. coli. The Recombinant Human IGFBP-rp1/IGFBP-7 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001544
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 2mM DTT, 1mM EDTA
Gene ID (Entrez) 3490
Molecular Weight (g/mol) 28.8kDa
Name IGFBP-rp1/IGFBP-7 Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen IGFBP7, 27-282aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSSSSDTCG PCEPASCPPL PPLGCLLGET RDACGCCPMC ARGEGEPCGG GGAGRGYCAP GMECVKSRKR RKGKAGAAAG GPGVSGVCVC KSRYPVCGSD GTTYPSGCQL RAASQRAESR GEKAITQVSK GTCEQGPSIV TPPKDIWNVT GAQVYLSCEV IGIPTPVLIW NKVKRGHYGV QRTELLPGDR DNLAIQTRGG PEKHEVTGWV LVSPLSKEDA GEYECHASNS QGQASASAKI TVVDALHEIP VKKGEGAEL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.