Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-32 His Protein
SDP

Catalog No. NBC125853 Shop All R&D Systems Products
Change view
Click to view available options
0.1mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBC125853 0.1mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC125853 Supplier Novus Biologicals™ Supplier No. NBC125853
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-131 of Human IL-32 The Recombinant Human IL-32 Protein is derived from E. coli. The Recombinant Human IL-32 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001012651
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 9235
Molecular Weight (g/mol) 19.1kDa
Name IL-32 Protein
Purification Method Affinity purified
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMCF PKVLSDDMKK LKARMHQAIE RFYDKMQNAE SGRGQVMSSL AELEDDFKEG YLETVAAYYE EQHPELTPLL EKERDGLRCR GNRSPVPDVE DPATEEPGES FCDKSYGAPR GDKEELTPQK CSEPQSSK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.