Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SF20/MYDGF His Protein
SDP

Catalog No. NBC122603 Shop All R&D Systems Products
Change view
Click to view available options
:
0.1mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBC122603 0.1mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC122603 Supplier Novus Biologicals™ Supplier No. NBC122603
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 33-173 of Human IL25 The Recombinant Human IL25 Protein is derived from E. coli. The Recombinant Human IL25 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_061980
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 20% glycerol
Gene ID (Entrez) 56005
Molecular Weight (g/mol) 18kDa
Name IL25 Protein
Purification Method Protein
Immunogen MGSSHHHHHH SSGLVPRGSH MSEPTTVAFD VRPGGVVHSF SHNVGPGDKY TCMFTYASQG GTNEQWQMSL GTSEDHQHFT CTIWRPQGKS YLYFTQFKAE VRGAEIEYAM AYSKAAFERE SDVPLKTEEF EVTKTAVAHR PGAFKAELSK LVIVAKASRT EL
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Gene Alias C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Protein SF20/MYDGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.