Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human JAZF1 His Protein
SDP

Catalog No. NBP223155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223155 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223155 Supplier Novus Biologicals™ Supplier No. NBP223155
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-243 of Human JAZF1 The Recombinant Human JAZF1 Protein is derived from E. coli. The Recombinant Human JAZF1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_778231
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 221895
Molecular Weight (g/mol) 29.5kDa
Name JAZF1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen JAZF1, 1-243aa, Human His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTGIAAA SFFSNTCRFG GCGLHFPTLA DLIEHIEDNH IDTDPRVLEK QELQQPTYVA LSYINRFMTD AARREQESLK KKIQPKLSLT LSSSVSRGNV STPPRHSSGS LTPPVTPPIT PSSSFRSSTP TGSEYDEEEV DYEESDSDES WTTESAISSE AILSSMCMNG GEEKPFACPV PGCKKRYKNV NGIKYHAKNG HRTQIRVRKP FKCRCGKSYK TAQGLRHHTI NFHPPVSAEI IRKMQQ
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.