Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human JDP2 His Protein
SDP

Catalog No. NBP223158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223158 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223158 Supplier Novus Biologicals™ Supplier No. NBP223158
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-163 of Human His-tag JDP2 The Recombinant Human JDP2 Protein is derived from E. coli. The Recombinant Human JDP2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_569736
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 1mM DTT, 1mM EDTA, 250mM imidazole
Gene ID (Entrez) 122953
Molecular Weight (g/mol) 21.2kDa
Name JDP2 Protein
Purification Method SDS-PAGE
Quantity 0.01 mg
Source E. Coli
Immunogen JDP2, 1-163aa, Human His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMMPGQI PDPSVTTGSL PGLGPLTGLP SSALTVEELK YADIRNLGAM IAPLHFLEVK LGKRPQPVKS ELDEEEERRK RRREKNKVAA ARCRNKKKER TEFLQRESER LELMNAELKT QIEELKQERQ QLILMLNRHR PTCIVRTDSV KTPESEGNPL LEQLEKK
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.