Learn More
Novus Biologicals™ Human Kidney Tissue Lysate (Female Adult Normal, Age:37)

Specifications
Specifications
| Tissue | Kidney |
| Quantity | 0.1 mg |
| Test Specificity | Sex: Female; Age: 37; Location: Right kidney; Diagnosis: Clear cell carcinoma; Grade: 1; Stage: NA |
| Tissue Condition | Normal |
| Host Species | Human |
| Concentration | 1 mg/mL |
| Product Type | Human Kidney Tissue Lysate (Adult Normal) |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Life Stage | Adult |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.