Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human LIN7C His Protein
SDP

Catalog No. NBP223199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223199 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223199 Supplier Novus Biologicals™ Supplier No. NBP223199
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-197 of Human LIN7C The Recombinant Human LIN7C Protein is derived from E. coli. The Recombinant Human LIN7C Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_060832
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 30% glycerol
Gene ID (Entrez) 55327
Molecular Weight (g/mol) 24.2kDa
Name LIN7C Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen LIN7C, 1-197 aa, Human, His tag, E.coli MGCSHHHHHH SSGLVPRGSH MGSMAALGEP VRLERDICRA IELLEKLQRS GEVPPQKLQA LQRVLQSEFC NAVREVYEHV YETVDISSSP EVRANATAKA TVAAFAASEG HSHPRVVELP KTEEGLGFNI MGGKEQNSPI YISRIIPGGI ADRHGGLKRG DQLLSVNGVS VEGEHHEKAV ELLKAAQGKV KLVVRYTPKV LEEMESRFEK MRSAKRRQQT
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.