Learn More
Novus Biologicals™ Human Liver Tissue Lysate (Adult)

Specifications
Specifications
| Tissue | Liver |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | HEPES (pH 7.9), KCl, Sucrose, Sodium deoxycholate, and NP-40, MagCl2, EDTA, Glycerol, and a cocktail of protease inhibitors |
| Host Species | Human |
| Concentration | 2 mg/mL |
| Product Type | Human Liver Tissue Lysate (Adult) |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
| Life Stage | Adult |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.