Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human LMW-PTP/ACP1 His Protein
SDP

Catalog No. NBC121279 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC121279 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC121279 Supplier Novus Biologicals™ Supplier No. NBC121279
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Specific activity is > 15,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.

Specifications

Accession Number NP_009030
Concentration 1 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20 mM MES (pH 6.0), 0.1 mM PMSF, 2 mM EDTA, 10% glycerol
Gene ID (Entrez) 52
Molecular Weight (g/mol) M.W. (theoretical): 20.1 kDa
Name LMW-PTP/ACP1 Protein
Purification Method Affinity purified
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MAEQATKSVL FVCLGNICRS PIAEAVFRKL VTDQNISENW VIDSGAVSDW NVGRSPDPRA VSCLRNHGIH TAHKARQITK EDFATFDYIL CMDESNLRDL NRKSNQVKTC KAKIELLGSY DPQKQLIIED PYYGNDSDFE TVYQQCVRCC RAFLEKAH
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Endotoxin Concentration <1.0 EU / 1 μg of protein (determined by LAL method)
Gene Symbol ACP1
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Protein LMW-PTP/ACP1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.