Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MAT1A His Protein
SDP

Catalog No. NBP140408 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140408 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140408 Supplier Novus Biologicals™ Supplier No. NBP140408
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-395 of Human MAT1A The Recombinant Human MAT1A Protein is derived from E. coli. The Recombinant Human MAT1A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q00266
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 0.1 M NaCl, 10% glycerol
Gene ID (Entrez) 4143
Molecular Weight (g/mol) 45.6kDa
Name MAT1A Protein
Purification Method Protein
Quantity 0.05 mg
Source E. Coli
Immunogen MAT1A, 1-395aa, Human, His tag, E.coli (NP_000420).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.