Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MEOX 2 His Protein
SDP

Catalog No. NBP223228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223228 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223228 Supplier Novus Biologicals™ Supplier No. NBP223228
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 188-304 of Human MEOX 2 The Recombinant Human MEOX 2 Protein is derived from E. coli. The Recombinant Human MEOX 2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005915
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 40% glycerol, 2mM DTT
Gene ID (Entrez) 4223
Molecular Weight (g/mol) 15.9kDa
Name MEOX 2 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen MEOX2, 188-304aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSRKERTAF TKEQIRELEA EFAHHNYLTR LRRYEIAVNL DLTERQVKVW FQNRRMKWKR VKGGQQGAAA REKELVNVKK GTLLPSELSG IGAATLQQTG DSIANEDSHD SDHSSEHAHL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.