Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MyD88 His Protein
SDP

Catalog No. NBP223255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223255 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223255 Supplier Novus Biologicals™ Supplier No. NBP223255
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-309 of Human MyD88 The Recombinant Human MyD88 Protein is derived from E. coli. The Recombinant Human MyD88 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002459
Concentration .5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 4615
Molecular Weight (g/mol) 38.7kDa
Name MyD88 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen MYD88, 1-309aa, Human, His tag, E.coli MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMRPD RAEAPGPPAM AAGGPGAGSA APVSSTSSLP LAALNMRVRR RLSLFLNVRT QVAADWTALA EEMDFEYLEI RQLETQADPT GRLLDAWQGR PGASVGRLLE LLTKLGRDDV LLELGPSIEE DCQKYILKQQ QEEAEKPLQV AAVDSSVPRT AELAGITTLD DPLGHMPERF DAFICYCPSD IQFVQEMIRQ LEQTNYRLKL CVSDRDVLPG TCVWSIASEL IEKRCRRMVV VVSDDYLQSK ECDFQTKFAL SLSPGAHQKR LIPIKYKAMK KEFPSILRFI TVCDYTNPCT KSWFWTRLAK ALSLP
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.