Learn More
Description
Specifications
Specifications
| Accession Number | NP_000423 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 5mM CaCl2 |
| Gene ID (Entrez) | 4633 |
| Molecular Weight (g/mol) | 20.9kDa |
| Name | Myosin Light Chain 2 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHHSSGLVPRGSHMAPKKAKKRAGGANSNVFSMFEQTQIQEFKEAFTIMDQNRDGFIDKNDLRDTFAALGRVNVKNEEIDEMIKEAPGPINFTVFLTMFGEKLKGADPEETILNAFKVFDPEGKGVLKADYVREMLTTQAERFSKEEVDQMFAAFPPDVTGNLDYKNLVHIITHGEEKD |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <1 EU per 1ug of protein (determined by LAL method) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
