Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NCK2 His Protein
SDP

Catalog No. NBP223272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223272 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223272 Supplier Novus Biologicals™ Supplier No. NBP223272
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-380 of Human NCK2 The Recombinant Human NCK2 Protein is derived from E. coli. The Recombinant Human NCK2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003572
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 8440
Molecular Weight (g/mol) 45.3kDa
Name NCK2 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen NCK2, 1-380aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTEEVIV IAKWDYTAQQ DQELDIKKNE RLWLLDDSKT WWRVRNAANR TGYVPSNYVE RKNSLKKGSL VKNLKDTLGL GKTRRKTSAR DASPTPSTDA EYPANGSGAD RIYDLNIPAF VKFAYVAERE DELSLVKGSR VTVMEKCSDG WWRGSYNGQI GWFPSNYVLE EVDEAAAESP SFLSLRKGAS LSNGQGSRVL HVVQTLYPFS SVTEEELNFE KGETMEVIEK PENDPEWWKC KNARGQVGLV PKNYVVVLSD GPALHPAHAP QISYTGPSSS GRFAGREWYY GNVTRHQAEC ALNERGVEGD FLIRDSESSP SDFSVSLKAS GKNKHFKVQL VDNVYCIGQR RFHTMDELVE HYKKAPIFTS EHGEKLYLVR ALQ
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.