Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NEK7 His Protein
SDP

Catalog No. NBP223287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223287 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223287 Supplier Novus Biologicals™ Supplier No. NBP223287
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-302 of Human NEK7 The Recombinant Human NEK7 Protein is derived from E. coli. The Recombinant Human NEK7 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_598001
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol
Gene ID (Entrez) 140609
Molecular Weight (g/mol) 37kDa
Name NEK7 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen NEK7, 1-302aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMDEQSQ GMQGPPVPQF QPQKALRPDM GYNTLANFRI EKKIGRGQFS EVYRAACLLD GVPVALKKVQ IFDLMDAKAR ADCIKEIDLL KQLNHPNVIK YYASFIEDNE LNIVLELADA GDLSRMIKHF KKQKRLIPER TVWKYFVQLC SALEHMHSRR VMHRDIKPAN VFITATGVVK LGDLGLGRFF SSKTTAAHSL VGTPYYMSPE RIHENGYNFK SDIWSLGCLL YEMAALQSPF YGDKMNLYSL CKKIEQCDYP PLPSDHYSEE LRQLVNMCIN PDPEKRPDVT YVYDVAKRMH ACTASS
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Protein Kinase
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.