Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NUDT4 His Protein
SDP

Catalog No. NBP223304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223304 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223304 Supplier Novus Biologicals™ Supplier No. NBP223304
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-180 of Human NUDT4 The Recombinant Human NUDT4 Protein is derived from E. coli. The Recombinant Human NUDT4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_061967
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 11163
Molecular Weight (g/mol) 22.7kDa
Name NUDT4 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen NUDT4, 1-180aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMMKFKPN QTRTYDREGF KKRAACLCFR SEQEDEVLLV SSSRYPDQWI VPGGGMEPEE EPGGAAVREV YEEAGVKGKL GRLLGIFENQ DRKHRTYVYV LTVTEILEDW EDSVNIGRKR EWFKVEDAIK VLQCHKPVHA EYLEKLKLGC SPANGNSTVP SLPDNNALFV TAAQTSGLPS SVR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.