Learn More
Description
Specifications
Specifications
| Accession Number | NP_002525 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT |
| Gene ID (Entrez) | 4938 |
| Molecular Weight (g/mol) | 43.9kDa |
| Name | OAS1 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MMDLRNTPAK SLDKFIEDYL LPDTCFRMQI NHAIDIICGF LKERCFRGSS YPVCVSKVVK GGSSGKGTTL RGRSDADLVV FLSPLTTFQD QLNRRGEFIQ EIRRQLEACQ RERAFSVKFE VQAPRWGNPR ALSFVLSSLQ LGEGVEFDVL PAFDALGQLT GSYKPNPQIY VKLIEECTDL QKEGEFSTCF TELQRDFLKQ RPTKLKSLIR LVKHWYQNCK KKLGKLPPQY ALELLTVYAW ERGSMKTHFN TAQGFRTVLE LVINYQQLCI YWTKYYDFKN PIIEKYLRRQ LTKPRPVILD PADPTGNLGG GDPKGWRQLA QEAEAWLNYP CFKNWDGSPV SSWILLVRPP ASSLPFIPAP LHEA |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
