Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human OXSM His Protein
SDP

Catalog No. NBP222661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222661 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222661 Supplier Novus Biologicals™ Supplier No. NBP222661
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 28-459 of Human OXSM The Recombinant Human OXSM Protein is derived from E. coli. The Recombinant Human OXSM Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_060367
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 54995
Molecular Weight (g/mol) 48.1kDa
Name OXSM Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen OXSM, 28-459aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSKRKFFGT VPISRLHRRV VITGIGLVTP LGVGTHLVWD RLIGGESGIV SLVGEEYKSI PCSVAAYVPR GSDEGQFNEQ NFVSKSDIKS MSSPTIMAIG AAELAMKDSG WHPQSEADQV ATGVAIGMGM IPLEVVSETA LNFQTKGYNK VSPFFVPKIL VNMAAGQVSI RYKLKGPNHA VSTACTTGAH AVGDSFRFIA HGDADVMVAG GTDSCISPLS LAGFSRARAL STNSDPKLAC RPFHPKRDGF VMGEGAAVLV LEEYEHAVQR RARIYAEVLG YGLSGDAGHI TAPDPEGEGA LRCMAAALKD AGVQPEEISY INAHATSTPL GDAAENKAIK HLFKDHAYAL AVSSTKGATG HLLGAAGAVE AAFTTLACYY QKLPPTLNLD CSEPEFDLNY VPLKAQEWKT EKRFIGLTNS FGFGGTNATL CIAGL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.