Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PAK4 His Protein
SDP

Catalog No. NBC118517 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118517 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118517 Supplier Novus Biologicals™ Supplier No. NBC118517
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-591 of Human PAK4 The Recombinant Human PAK4 Protein is derived from E. coli. The Recombinant Human PAK4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number O96013
For Use With (Application) ELISA, SDS-PAGE
Formulation 25mM HEPES buffer (pH 7.5) 150mM NaCl, 5mM MgCl2, 30% glycerol
Gene ID (Entrez) 10298
Molecular Weight (g/mol) 68.3kDa
Name PAK4 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen Human PAK4, 1-591aa (NP_001014831 ).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.